Principaux documents
23628
Supelco
Connecteur GalsSeal pour colonnes capillaires
pkg of 25 ea
About This Item
Code UNSPSC :
41115700
Conditionnement
pkg of 25 ea
Technique(s)
gas chromatography (GC): suitable
Description générale
GlasSeal connectors quickly connect fused silica tubing of the same or different diameter with no tools and no leaks. Use them to connect a guard column or transfer line, repair a broken column, or connect two columns (same or different phases).
A Y connector splits a sample to two columns or a column effluent to two detectors. All of these connectors are silanized for an inert inside surface. Choose borosilicate glass or fused silica. Use with our 0.25mm-0.53mm I.D. tubing.
Application
DB5 MS column was joined to the mass spectrometer end to reduce column bleed entering the mass spectrometer using GlasSeal capillary column connector during chiral analysis. It was used to connect three fused silica columns during analyses of pheromone using GC.
Autres remarques
We offer a variety of chromatography accessories including analytical syringes
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
R R Heath et al.
Journal of chemical ecology, 19(10), 2395-2410 (1993-10-01)
A bioassay system was developed that permits the testing of various substrates for biological activity in a flight tunnel, while simultaneously collecting a portion of the volatiles from the attractive source for subsequent chemical identification and quantification. Bioassays of the
A T Fisk et al.
Archives of environmental contamination and toxicology, 42(1), 118-126 (2001-11-14)
To examine the influence of diet and age on organochlorine contaminant (OC) concentrations in two closely related ringed seal (Phoca hispida) populations enantiomeric fractions (EFs) of chiral contaminants and stable isotopes of nitrogen (delta15N) and carbon (delta13C) were measured along