Principaux documents
Z756695
Scienceware® 50 mL tube holder
non-grip style, round white polystyrene
Synonyme(s) :
centrifuge tube holder, tube rack
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.22
Matériaux
round white polystyrene
Caractéristiques
Holds 1 x 50 mL tube
Conditionnement
bag of 5 ea
Fabricant/nom de marque
Bel-Art F18795-0000
Description générale
Leave both hands free for pipetting and other critical tasks. The Grip Style gently grips the tube and moves with the tube when it is lifted from the bench top.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP