Principaux documents
Z709638
Sartorius mLINE® mechanical pipettor
12 channel, volume 0.5-10 μL
Synonyme(s) :
Biohit pipettes, pipettor
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Caractéristiques
12-channel
electronic: no
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Sartorius 725220
Plage de mesure du volume
0.5-10 μL
Volume
10 μL
Description générale
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
  • Extremely light pipetting and tip ejection
  • Unique safety features
  • Stepless volume adjustment and patented volume locking system
  • Thermal insulation of internal components to improve accuracy
  • Full autoclavability without disassembling
  • Easy cleaning, maintenance and calibration
  • ID tag and color-coded caps
  • Safe-Cone Filters and unique ejection
  • Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Informations légales
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy