Key Documents
Z331457
Lenz® round bottom flasks
capacity 100 mL, joint: ST/NS 29/32
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass 3.3
round bottom flask
single-neck
Joint
joint: ST/NS29/32
Agence
according to DIN 12 348
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Lenz 3.0029.37
Capacité
100 mL
Informations légales
Lenz is a registered trademark of Lenz GmbH