Key Documents
MTCR2781P
MTC Bio FlipTop Freezer Box With Hinged Lid
cardboard, purple, to hold, 81 x cryovials, pkg of 5 ea
Synonyme(s) :
cryo tube storage box, cryogenic storage, culture test tube, plastic tube
About This Item
Code UNSPSC :
41122806
Nomenclature NACRES :
NB.84
Matériaux
cardboard
purple
Caractéristiques
to hold 81 x cryovials
Conditionnement
pkg of 5 ea
Fabricant/nom de marque
MTC Bio R2781-P
L × l × H
133 mm × 133 mm × 51 mm
Description générale
FlipTop Carboard freezer box w/ attached hinged lid, 2 inch, 100-place, assorted colors, 5/pk
Recyclable cardboard, with a liquid resistant outer coating. The 81-place, 3 inch FlipTop box is suitable for tubes up to 13mm in diameter. The 100-place, 3 inch FlipTop box is suitable for tubes up to 12mm in diameter (accepts cryovials up to 4mL). Boxes are assembled complete with dividers. Each box has drain slots on the bottom and numbered grid markings in almost every conceivable location (top, bottom, sides).
Caractéristiques et avantages
  • Attached Hinged Lids
  • Foolproof identification
  • 5 1/4 x 5 1/4 x 2 inches
  • Standard size, for existing racks
  • Available in 6 colors
  • Individual or rainbow packs
  • For 81 or 100 tubes
  • 9×9 or 10×10 format
  • Alpha-numeric coding
  • Top, sides, and bottom
Informations légales
FlipTop is a trademark of MTC Bio
MTC is a trademark of MTC Bio