Principaux documents
MTCP5212200U
MTC Bio ProPette LE 12-Channel Adjustable Pipettor
capacity 20-200 μL
Synonyme(s) :
pipette
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.15
Caractéristiques
adjustable
manual
no. of channels 12
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio P5212-200U
Capacité
20-200 μL
Catégories apparentées
Description générale
Ideal for all 96-well applications, Propette LE Multi-Channel Pipettes are lightweight, rugged, and capable of standing up to the demands of everyday use in a busy laboratory environment.
ProPette LE 12 -Channel Pipette, 20 to 200µL
Caractéristiques et avantages
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (1.0µL-300µL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Informations légales
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio