MTCP1001A
MTC™ Bio Pure.Amp™ Aluminum Sealing Film
to fit standard PCR plates
Synonyme(s) :
PCR plate seal, aluminimum membrane, flow cytometry tube, sealing membrane
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.15
Produits recommandés
Conditionnement
pack of 100 ea
Fabricant/nom de marque
MTC™ Bio P1001-A
Description générale
Sealing film, Aluminum membrane, pk/100
PureAmp Sealing Membranes are compatible with all standard PCR plates. Pierceable aluminum membrane guaranteed to keep evaporation rates below 5%.
PureAmp Sealing Membranes are compatible with all standard PCR plates. Pierceable aluminum membrane guaranteed to keep evaporation rates below 5%.
Caractéristiques et avantages
- RNase, DNase, DNA, and PCR inhibitor free
- Evaporation rates ? 5%
- Inner packs of 10 membranes
- -80° to 120°C
Informations légales
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio