Merck
M4664
Nalgene® vacuum manifold
autoclavable
Synonyme(s) :
moisture extraction device, sample protection unit, Nalgene DS0345-0001, manifold, stainless steel vacuum manifold
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.24
UNSPSC Code:
41104900
Packaging:
pack of 1 ea
Manufacturer/tradename:
Nalgene DS0345-0001
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
feature
autoclavable
packaging
pack of 1 ea
manufacturer/tradename
Nalgene DS0345-0001
General description
Stainless steel with PTFE stopcocks. Designed with three vacuum ports each with a two-way valve and vent. Barbed fitting accepts 3/8 in. (9.5 mm) I.D. tubing.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries