GBL103259
Grace Bio-Labs CultureWell™ removal tool
for removable chambered coverglass
Synonyme(s) :
chambered coverglass
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.22
Produits recommandés
Matériaux
acrylic
blue
Stérilité
non-sterile
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Grace Bio-Labs 103259
Taille externe
53 mm × 100 mm
Description générale
Designed to separate the chambered upper structure from the coverglass and leave a removable gasket on the coverglass.
Informations légales
Culturewell is a trademark of Grace Bio-Labs, Inc.