Principaux documents
EP5409712004
Rotor for Eppendorf® 5427R Centrifuge
FA-45-48-11, with rotor lid, to hold, 48 x 1.5/2.0 mL tubes
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 48 x 1.5/2.0 mL tubes
fixed angle rotor
with rotor lid
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5409712004 (Global)
Paramètres
12,700 rpm
18,210 × g max. RCF
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5427R
Description générale
Rotors to fit Eppendorf 5427 R refrigerated centrifuge.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG