Principaux documents
EP4960824003
Sigma-Aldrich
Eppendorf® Discharge Tube Spiral
for Eppendorf® Varispenser® and Varispenser® plus 2.5 mL/5 mL/10 mL, L 80 cm, volume 2.5-10 mL
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.02
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 4960824003 (Global)
Longueur
80 cm
Volume
2.5-10 mL
Application
Liquid handling
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Varispenser is a registered trademark of Eppendorf AG