Principaux documents
EP0030128583
Eppendorf® Twin tec PCR plates, 96 wells
yellow plate, conical bottom yellow wells, semi-skirted, pkg of 25 ea
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
Matériaux:
conical bottom yellow wells
polycarbonate
yellow plate
Taille:
96 wells
Volume maximum des puits:
250 μm
Volume de travail:
250 μL
Stérilité:
non-sterile
Caractéristiques:
skirt (Semi skirted)
Matériaux
conical bottom yellow wells
polycarbonate
yellow plate
Stérilité
non-sterile
Caractéristiques
skirt (Semi skirted)
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030128583
Technique(s)
PCR: suitable
Taille
96 wells
Volume maximum des puits
250 μm
Volume de travail
250 μL
Description générale
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG