Principaux documents
CLS6795600D
Corning® Digital Hotplates
AC/DC input 120 V AC, US 3-pin plug, L × W 10 nL × 10 in.
Synonyme(s) :
stirrers
About This Item
Code UNSPSC :
41102406
Nomenclature NACRES :
NB.76
Matériaux
ceramic plate
Conditionnement
case of 1
Fabricant/nom de marque
Corning 6795-600D
Disponibilité
not available in EU
Paramètres
5-550 °C temp. range
L × l
10 nL × 10 in.
Catégories apparentées
Description générale
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Microprocessor maintains consistent and repeatable temperature settings.
  • Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
  • Separate temperature sensor provides power cut off if unit overheats
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
  • 2 year warranty
Informations légales
Corning is a registered trademark of Corning, Inc.