CLS6628
Corning® Step-R Repeating Pipettor Syringe Tips
volume (0.05 mL), sterile
Synonyme(s) :
pipet tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
Produits recommandés
Stérilité
sterile
Conditionnement
pack of 100 ea
Fabricant/nom de marque
Corning 6628
Volume
(0.05 mL)
Description générale
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Informations légales
Corning is a registered trademark of Corning, Inc.