Principaux documents
CLS480126
Corning® LSE single block, for Corning LSE single block digital dry baths
single block, 12 x 17 mm tubes, 1/cs
Synonyme(s) :
equipment replacement accessory
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NC.21
Caractéristiques
Holds 12 x 17 mm tubes
Conditionnement
pack of 1
case of 2
Fabricant/nom de marque
Corning 480126
Adéquation
suitable for (use with Corning LSE digital dry bath heater, single block)
Description générale
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
Informations légales
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.