Principaux documents
CLS352196
Corning® Falcon® Centrifuge Tubes
capacity 15 mL, conical polypropylene, cap, dome seal screw, bagged, sterile
Synonyme(s) :
culture tube
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.13
Matériaux
conical polypropylene
Stérilité
sterile
Caractéristiques
cap dome seal screw
Fabricant/nom de marque
Corning 352196
Capacité
15 mL
Application
Liquid Handling
Informations légales
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.