Principaux documents
Z757993
Q-Tube gas purging set
pkg of 1 ea
Synonyme(s) :
Q-Tube gas purging set, pressure reactor, q-tube pressure reactor
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Q Labtech
Catégories apparentées
Description générale
This set can be operated safely and accurately with high reproducibility with a wide range of gases, including oxygen, nitrogen, argon, hydrogen, deuterium, carbon monoxide, and carbon dioxide.
The set contains all items needed for immediate use:
  • 316 stainless steel adapter and matched PTFE sleeve
  • 1 pressure gauge, 300 psi, 2 valves
  • 1 pressure tube, 35 mL
  • 20 septa
  • All fittings for connecting to a gas regulator
Informations légales
Q-Tube is a trademark of Q Labtech LLC