Principaux documents
Z684872
Quick-release clamps
with 5/8 x 12 in. extension, for 150 mm (6 in.) Schott flange
Synonyme(s) :
clamps, quick release clamp
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.43
Matériaux
stainless steel
Stérilité
non-sterile
Conditionnement
case of 1
Fabricant/nom de marque
Ace Glass 6517-56
Description générale
SS clamps for use with reaction flasks and heads with a 100 mm Schott flange. The extension rod can be clamped to the support frame and helps stabilize the reaction assembly.