Principaux documents
Z682446
Sigma-Aldrich
Ace vapor duct tube, for condenser "V" assembly
joint: ST/NS 24/40, L 45 mm, fits Büchi® models R-210/R-215
Synonyme(s) :
Ace vapor duct tube, for condenser "V" assembly
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Büchi equivalent, 48067
Longueur
45 mm
Catégories apparentées
Description générale
The vapor duct tube accessories are for Büchi condenser "V" assemblies, and will fit either Büchi Rotavapor model R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG