Z552682
Aldrich® Thermometer Adapter
straight, male joint: ST/NS 14/20, GL 14 thread
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
male joint: ST/NS14/20
Raccords
GL 14 thread
Description générale
Has PTFE faced silicone washer and open-top cap.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC