Principaux documents
Z549622
Sigma-Aldrich
Stopcock Adapter
PTFE side-port stopcock, joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Matériaux
PTFE side-port stopcock
Joint
joint: ST/NS24/40
Catégories apparentées
Description générale
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.