Key Documents
Z530255
Sigma-Aldrich
Aldrich® vacuum-transfer adapter
joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS14/20
Taille du robinet d'arrêt
2 mm
Description générale
PTFE stopcocks. For transfer of hazardous and sensitive reagents.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC