Key Documents
Z516473
Sigma-Aldrich
Aldrich® 5 mm NMR tube evacuation manifold
1 port, L 100 mm
Synonyme(s) :
nmr tube, nmr tubes, tube manifold
About This Item
Code UNSPSC :
41121800
eCl@ss :
32129209
Nomenclature NACRES :
SB.32
Longueur
100 mm
Description générale
Designed to evacuate NMR tubes that have a J Young PTFE valve. Once the NMR tube has been evacuated in the manifold, close the valve and place into the NMR spectrometer. Manifolds have 0-10 mm bore PTFE valves. The multiple evacuation station units allow removal of one NMR tube while the remaining tube(s) stay under vacuum.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC