Principaux documents
Z513539
Sigma-Aldrich
Aldrich® mini gas regulator
brass body
Synonyme(s) :
Small Gas Regulator
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Matériaux
brass body
Description générale
These high-purity, single-stage, single-gauge regulators have a 316 SS diaphragm for accurate, stable pressure control of Aldrich liquefied gases. Delivery pressure: 1-50 psi, 1/4 in. NPTF inlet, hose barb outlet.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC