Principaux documents
Z420220
Sigma-Aldrich
Spinbar® disposable magnetic stirring bar
size 1 in. × 5/16 in.
Synonyme(s) :
Stir Bar, Stirbar, Stirring Bar
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
PTFE-coated
polygon PTFE-coated
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
Bel-Art F37112-0001
Taille
1 in. × 5/16 in.
Description générale
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.
Informations légales
Spinbar is a registered trademark of SP Scienceware, a division of SP