Key Documents
Z304832
Quickfit® drying tube
joint: ST/NS 19/26
Synonyme(s) :
MF23/2
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
colorless tube
glass tube
Joint
joint: ST/NS19/26
Stérilité
non-sterile
Caractéristiques
graduations: no
neck thread PUSH FIT (19/26 cone size)
Fabricant/nom de marque
Quickfit MF 23/2
Disponibilité
not available in USA and Canada
Type de liaison
non-treated surface
Description générale
Angled. Retain desiccant with plug of glass wool.
Informations légales
Quickfit is a registered trademark of SciLabware Limited