Documents
Z244120
Sigma-Aldrich
Sample cylinder plug
carbon steel
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Matériaux
carbon steel
Description générale
The 1/4 in. NPTM square head plug fits all Sure/Pac sample cylinder sizes.