Z225282
Precision crucible tongs
L 20 in. (508 mm)
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
Produits recommandés
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Longueur
20 in. (508 mm)
Description générale
Stainless steel tongs with serrated tapered tips, which meet precisely when closed.