Z192732
Horn tip for 500 and 750W ultrasonic processor
for horn 1/2 in.
About This Item
Code UNSPSC :
41101500
Nomenclature NACRES :
NB.77
Produits recommandés
Fabricant/nom de marque
Sonics & Materials 630-0406
Description générale
Tips screw into end of threaded horn. Replace when cavitation erosion becomes excessive.