Key Documents
Z190357
Sigma-Aldrich
Support stand
rod diam. × L 5/8 in. × 29 in. , SS
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Tige, diam. × L
5/8 in. × 29 in. , SS
Description générale
Heavy steel base. Suitable for mounting stirring motors, etc.
Includes metal base and rod.
Les clients ont également consulté
Slide 1 of 1