Key Documents
Z186775
Gas-collecting bulb
polypropylene valve, capacity 75 cc, diam. 53 mm
About This Item
Code UNSPSC :
42272218
Nomenclature NACRES :
NB.51
Matériaux
polypropylene valve
Capacité
75 cc
Diamètre
53 mm
Description générale
For priming pumps, transfer lines, etc. Natural rubber with 3/8 in. O.D.