Key Documents
Z151432
Bransonic® ultrasonic cleaner accessories
Solid tray, stainless steel, tank capacity 2.8 L
About This Item
Code UNSPSC :
41101500
Nomenclature NACRES :
NB.77
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Branson 100-410-172
Capacité de la cuve
2.8 L
Description générale
Solid Stainless steel tray for use with Branson M series, model 2800 Utrasonic Bath. Dimensions 90 mm x 90 mm x 75 mm.
Informations légales
Bransonic is a registered trademark of Branson Ultrasonics Corp.