Principaux documents
Z122416
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 200 mm, joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/20
H totale
200 mm
Description générale
Condensers have a standard taper female joint and a lower male drip tip-joint.
Replacement part for Aldrich All-in-One Lab Glassware Kit.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC