Principaux documents
Z117129
Stainless steel 304 syringe needle
gauge 20, L 10 in.
Synonyme(s) :
Syringe needle
About This Item
Code UNSPSC :
41122002
eCl@ss :
32070500
Nomenclature NACRES :
NB.03
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cadence Science
Longueur
10 in.
Calibre
20
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing