Principaux documents
Z115355
Sigma-Aldrich
Spinbar® assortment
round PTFE-coated, pack of 6 ea
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
round PTFE-coated
Conditionnement
pack of 6 ea
Fabricant/nom de marque
Bel-Art F37161-0000
Description générale
All have removable pivot rings. 2 of each size. 1 × 5/16 in., 11/2 × 5/16 in., 2 × 5/16 in.
Informations légales
Spinbar is a registered trademark of SP Scienceware, a division of SP