Principaux documents
WHA10534320
Cartes Whatman® protein saver
903 Protein saver snap apart card, pkg of 100 ea
Synonyme(s) :
Whatman fta, Z694827, dbs cards, pcr blood
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
Whatman 10534320
Whatman Article No. 28416622 (US reference)
Description générale
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with four half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75-80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining cover is folded over the sample. This cover is imprinted with the universal biohazard symbol.
Caractéristiques et avantages
  • Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Les clients ont également consulté
Slide 1 of 1
1 of 1
Samira Asgari et al.
Nature, 582(7811), 234-239 (2020-06-06)
On average, Peruvian individuals are among the shortest in the world1. Here we show that Native American ancestry is associated with reduced height in an ethnically diverse group of Peruvian individuals, and identify a population-specific, missense variant in the FBN1
Protocoles
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.