Key Documents
TEM-FCF200NI50
Sigma-Aldrich
Formvar/Carbon Supported Nickel Grids
size 200 mesh, box of 50 ×
Synonyme(s) :
Formvar/Carbon 200 Mesh, Nckeli, 50pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50 ×
Taille
200 mesh
Description générale
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon