Principaux documents
TEM-CFT400CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 400 mesh, box of 50
Synonyme(s) :
CARBON THIN BAR SQUARE MESH - CU 400 MESH UL 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
400 mesh
Description générale
A 3-4 nm ultrathin film of pure carbon supported thin bar square mesh grid. Copper grid, 400 mesh. Carbon film show better electrical and heat conduction in the electron beam.