Key Documents
SYNS392440
Synthware cap design stopper
joint: ST/NS 24/40
Synonyme(s) :
Synthware S392440, glass stopper
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S392440
Description générale
For apparatus having an inner opening.
Informations légales
Synthware is a trademark of Kemtech America Inc.