SYNM531042
Synthware™ nitrogen/argon line manifold with PTFE stopcocks
stopcock bore 2 mm, Ports: 4, Hose Connections: left
Synonyme(s) :
Synthware M531042, vacuum manifold, gas manifold
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Produits recommandés
Fabricant/nom de marque
M531042
Alésage du robinet d'arrêt
2 mm
Catégories apparentées
Description générale
Synthware™ glass manifold featuring high vacuum hollow glass stopcocks, the smaller O.D. 17 mm front tube is designed for inert gases, while the larger O.D. 22 mm back tube is for vacuum, which has a 35/20 socket on the left-hand side. Hose connections (10 mm) are located front-left-right.
Informations légales
Synthware is a trademark of Kemtech America Inc.