Principaux documents
SYNF541100
Synthware reaction flask with high vacuum valve
100 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonyme(s) :
Synthware F541100, Schlenk flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
single-neck (high vaccum valve)
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F541100
Capacité
100 mL
100 mL
Taille de la vanne
0-4 mm
Description générale
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Viton® O-ring on shaft.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company