Principaux documents
SYNF43401L
Synthware three neck round bottom flask with angled side necks
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonyme(s) :
Synthware F43401L, 3-neck round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
three-neck (angled side necks)
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F43401L
Capacité
1,000 mL
1000 mL
1000 mL
Description générale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.