Principaux documents
SYNF110350
Synthware Dewar flask, tall form with metal housing
flask capacity 350 mL, tall form with metal housing
Synonyme(s) :
Synthware F110350
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
aluminum flask
Joint
Joints neck (70)
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F110350
D.I. × épaisseur
7 cm × 12.5 cm
Contenance du flacon/de la fiole
350 mL
Description générale
Dewar flask with weighted metal base for stability and with a full-length aluminum housing which extends to the top rim for added protection.
Informations légales
Synthware is a trademark of Kemtech America Inc.