Key Documents
SYNC574200
Synthware water jacketed distillation column with removable hose connections
joint: ST/NS 24/40, jacket H 200 mm
Synonyme(s) :
Synthware C574200, glass distilling column
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C574200
H de l'enveloppe
200 mm
Description générale
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Informations légales
Synthware is a trademark of Kemtech America Inc.