Principaux documents
SYNA262414
Synthware Kjeldahl distilling trap
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 14/20
Synonyme(s) :
Synthware A262414, Distilling trap, Kjeldahl trap
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS14/20
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A262414
Capacité
100 mL
Description générale
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Articles
Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.
Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.
Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.
Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.