Principaux documents
SLW1516/01RED
Pyrex® Media-Lab Bottles, with high temperature cap and pouring ring, with printed trace code
capacity 25 mL
Synonyme(s) :
Pyrex, SciLabware 1516/01RED
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Capacité:
25 mL
Diam. × H:
36 mm × 70 mm
Matériaux:
PTFE seal
red polyester cap (high temperature thermoplastic)
white graduation
Caractéristiques:
graduations
with cap
with pouring ring
with printed trace code
Conditionnement:
pack of 10 ea
Matériaux
PTFE seal
red polyester cap (high temperature thermoplastic)
white graduation
Joint
Joints threaded neck (GL25)
Caractéristiques
graduations
with cap
with pouring ring
with printed trace code
Conditionnement
pack of 10 ea
Fabricant/nom de marque
SciLabware 1516/01RED
Capacité
25 mL
Diam. × H
36 mm × 70 mm
Description générale
  • Bottles complete with thermoplastic polyester cap, PTFE sealing disc and pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Withstands hot air sterilisation up to 180 deg. C
  • PTFE sealing disc resists chemical attack
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Informations légales
PYREX is a registered trademark of Corning, Inc.