Key Documents
CLS999815
Corning® phenolic cap, PTFE liner
15-415 thread
Synonyme(s) :
Corning cellstack cell culture chambers, black phenolic, phenolic caps, screw caps, tissue culture chambers, Corning® 9998
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.43
Matériaux
PTFE liner
black phenolic cap
Fabricant/nom de marque
Corning 9998-15
Hauteur
~16 mm
Raccords
15-415 thread
Description générale
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Informations légales
Corning is a registered trademark of Corning, Inc.