Principaux documents
BR782150
BRAND® lids for microplates BRAND®plates
plate size 96 wells, with condensation rings
Synonyme(s) :
Accessories, BRANDplates®, Lids, Microplates
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux:
polystyrene
Taille de la plaque/du plateau:
96 wells
Matériaux
polystyrene
Conditionnement
pack of 100 ea
Fabricant/nom de marque
BRAND 782150
Taille de la plaque/du plateau
96 wells
Description générale
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG