Key Documents
BR780702
BRAND® micro tube with attached screw cap and sealing cone
capacity 1.5 mL, conical bottom, non-sterile
Synonyme(s) :
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes
About This Item
Code UNSPSC :
41121702
Nomenclature NACRES :
NB.11
Matériaux
colorless cap
colorless tube
conical bottom
polyethylene cap
polypropylene tube
screw top cap
Stérilité
non-sterile
Caractéristiques
graduations: no
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
BRAND 780702
Capacité
1.5 mL
Diam. du tube
13 mm
Description générale
BRAND reaction tubes with attached cover (PE) reduce the danger of contamination or mistakes due to mixing up covers. The sealing cone of the cover lies seamlessly against the interior of the tube, preventing the contents from coming into contact with the thread.Application: -90 °C to +100 °CAttached covers are suitable for colored cover inserts. BIO-CERT® PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Informations légales
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG