Principaux documents
BR51422
BRAND® ground joint sleeve
PTFE, joint: 29/32, thin-walled
Synonyme(s) :
PTFE
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Matériaux
PTFE
Joint
joint: 29/32
Caractéristiques
thin-walled
Fabricant/nom de marque
BRAND 51422
Description générale
Made from PTFE, this greaseless seal ensures that samples remain uncontaminated. It offers exceptional chemical resistance across a wide temperature range, from -200 to +260 °C. The thin-walled sleeves, measuring just 0.05 mm thick, fit snugly without adding bulk. The joints are designed for a perfect fit without a protruding rim, simplifying the use of joint clips. This versatile product is suitable for a wide range of applications, including vacuum conditions down to 0.1 mbar, making it ideal for use with rotary evaporators.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG